Ammonium carbonate, Puriss., meets analytical specification of NF, Ph. Franc., FCC, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Salt of hartshorn

11204-6X1KG 11204-2.5KG
BJ11204-1KGCS 344.24 USD
BJ11204-1KG BJ-11204-2.5KG
Ammonium carbonate, Puriss., meets analytical specification of NF, Ph. Franc., FCC, Fluka™
Ammonium carbonate
Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Vapor Density 2.7 (vs air)
Assay of NH₃ 30.0 - 34.0 %
Non-volatile matter Max. 0.05 %
Residue on ignition (SO₄) Max. 0.05 %
Arsenic (As) Max. 0.0003 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.0003 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR