Antimony(III) chloride ≥99.0% (RT), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Antimony trichloride

10774-500G
BJ10774-100GEA 157.59 USD
BJ10774-100G BJ10774-500G
Antimony(III) chloride ≥99.0% (RT), Puriss. p.a. ACS, Fluka™
Antimony (III) chloride
Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 283 °C (1.013 hPa)
Melting Pt: 73.4 °C
Density: 3.140 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insoluble in Chloroform Max. 0.05 %
Arsenic (As) Max. 200 ppm
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 5 ppm
Sulfate (SO₄) Max. 0.005 %
Vapor Pressure 1 mmHg ( 49 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR