Bismuth(III) nitrate basic 71-74% (Bi basis (calc. on dried substance)), Puriss., meets analytical specification of DAB, USP, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate

10334-1KG 10334-250G
BJ10334-1KGEA 382.43 USD
BJ10334-1KG BJ10334-250G
Bismuth(III) nitrate basic 71-74% (Bi basis (calc. on dried substance)), Puriss., meets analytical specification of DAB, USP, Fluka™
Bismuth (III) nitrate basic
Formula: Bi₅O(OH)₉(NO₃)₄
Melting Pt: >500 °C (dec.)
Density: 5.29 g/cm³ (22.8 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064844
CAS Number: 1304-85-4
EINECS: 215-136-8

Order Now

Specification Test Results

Assay of Bi (calculated to dried substances) 71 - 74 %
Loss on drying (105°C) Max. 3 %
Acidly reacting impurities complying
Ammonium (NH₄) Max. 0.01 %
Silver (Ag) Max. 0.0025 %
Arsenic (As) Max. 0.0002 %
Copper (Cu) Max. 0.005 %
Lead complying
Lead (Pb) Max. 0.002 %
Alkalis/earth alkalis Max. 0.5 %
Substances not precipitated by NH₄OH (SO₄) Max. 0.5 %
Carbonate (CO₃) complying
Chloride (Cl) Max. 0.02 %
Sulfate (SO₄) Max. 0.03 %
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR