Silver sulfate ≥99%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

10229-500G
BJ10229-500GEA 4063 USD
BJ10229-500G
Silver sulfate ≥99%, Puriss., Fluka™
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: Dec.
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Copper (Cu) Max. 0.005 %
Iron (Fe) Max. 0.005 %
Lead (Pb) Max. 0.005 %
Chloride (Cl) Max. 0.001 %
Nitrate (NO₃) Max. 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR