Chlorobenzene 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

101389-2.5L
BJ101389-2.5LEA 83.7 USD
BJ101389-2.5L
Chlorobenzene 99%, Reagent Grade
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 130-132 °C (1.013 hPa)
Melting Pt: -45 °C
Density: 1.107…1.109 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) min. 98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR