Ammonium sulfamate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Warning

09960-100G
BJ09960-100GEA 160.9 USD
BJ09960-100G
Ammonium sulfamate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
Ammonium sulphamate
Formula: H₆N₂O₃S
Melting Pt: 131…135 °C
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011429
CAS Number: 7773-06-0
EINECS: 231-871-7

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Melting point 131.5 - 133.5 °C
pH (5 %, 20°C) 5.0 - 6.0
Colour of the solution complying
Clearness of the solution complying
Sulfated ash Max. 0.02 %
Loss on drying (110°C) Max. 1 %
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Sulfate (SO₄) Max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR