Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

09874-100G
BJ09874-100GEA 307.03 USD
BJ09874-100G BJ09874-500G
Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™
Ammonium iodide
Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551
Density: 2.510 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 13,00531

Order Now

Specification Test Results

Assay 99.0 - 100.0 %
pH (5 %, 20°C) 4.5 - 6.5
Colour of the solution complying
Clearness of the solution complying
Residue on ignition (SO₄) Max. 0.02 %
Water insoluble matter Max. 0.005 %
Barium (Ba) Max. 20 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 2 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride and bromide (as Cl) Max. 50 ppm
Total S (as SO₄) Max. 20 ppm
Phosphate (PO₄) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR