Ammonium hydrogen sulfate ≥98.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ammonium bisulfate, Ammonium bisulphate

09848-1KG
BJ09848-1KGEA 151.8 USD
BJ09848-1KG
Ammonium hydrogen sulfate ≥98.0% (by titrimetric analysis), Purum p.a.
Ammonium hydrogen sulphate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: H₅NO₄S
MW: 115.11 g/mol
Boiling Pt: 350
Melting Pt: 138.85
Density: 1.790 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00051073
CAS Number: 7803-63-6
EINECS: 232-265-5
UN: 2506
ADR: 8 II
Merck Index: 12,00526

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR