Diammonium hydrogen citrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

09831-500G
BJ09831-500GEA 151.79 USD
BJ09831-500G
Diammonium hydrogen citrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
Density: 1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Residue on ignition Max. 0.01 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 500 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Oxalate (C₂O₄) Max. 500 ppm
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR