Ammonium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Warning

09725-100G
BJ09725-100GEA 268 USD
BJ09725-100G BJ09725-25G
Ammonium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Ammonium chloride

TraceSELECT™ Ultra and TraceSELECT


  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: NH₄Cl
Melting Pt: 338 °C (dec.)
Density: 1.536 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.01 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.05 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.1 ppm
Cesium (Cs) Max. 0.1 ppm
Copper (Cu) Max. 0.01 ppm
Dysprosium (Dy) Max. 0.01 ppm
Erbium (Er) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.05 ppm
Gadolinium (Gd) Max. 0.01 ppm
Germanium (Ge) Max. 0.05 ppm
Hafnium (Hf) Max. 0.01 ppm
Mercury (Hg) Max. 1 ppb
Holmium (Ho) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Iridium (Ir) Max. 0.01 ppm
Potassium (K) Max. 0.5 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.05 ppm
Lutetium (Lu) Max. 0.01 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.05 ppm
Sodium (Na) Max. 0.5 ppm
Niobium (nb) Max. 0.01 ppm
Neodymium (nd) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.01 ppm
Praseodymium (Pr) Max. 0.01 ppm
Platinum (Pt) Max. 0.1 ppm
Rubidium (Rb) Max. 0.01 ppm
Rhenium (Re) Max. 0.01 ppm
Rhodium (Rh) Max. 0.01 ppm
Ruthenium (Ru) Max. 0.01 ppm
Antimony (Sb) Max. 0.01 ppm
Scandium (Sc) Max. 0.05 ppm
Selenium (Se) Max. 0.05 ppm
Samarium (Sm) Max. 0.01 ppm
Tin (Sn) Max. 0.05 ppm
Strontium (Sr) Max. 0.1 ppm
Tantalum (Ta) Max. 0.01 ppm
Terbium (Tb) Max. 0.01 ppm
Tellurium (Te) Max. 0.01 ppm
Thorium (Th) Max. 0.01 ppm
Titanium (Ti) Max. 0.05 ppm
Thallium (Tl) Max. 0.01 ppm
Thulium (Tm) Max. 0.01 ppm
Uran (U) Max. 0.01 ppm
Vanadium (V) Max. 0.05 ppm
Tungsten (W) Max. 0.01 ppm
Yttrium (Y) Max. 0.05 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Zirconium (Zr) Max. 0.01 ppm
Nitrate (NO₃) Max. 5 ppm
Phosphate (PO₄) Max. 2 ppm
Sulfate (SO₄) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR