Ammonium iron (II) sulfate hexahydrate ≥99.0% (RT), Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

09720-1KG
BJ09720-1KGEA 170.04 USD
BJ09720-1KG
Ammonium iron (II) sulfate hexahydrate ≥99.0% (RT), Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: 100 °C (dec.)
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Assay (manganometric) Min. 99.0 %
pH (5 %, 25 °C) 3.0 - 5.0
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 20 ppm
Chromium (Cr) Max. 50 ppm
Copper (Cu) Max. 20 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 100 ppm
Manganese (Mn) Max. 500 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 200 ppm
Lead (Pb) Max. 10 ppm
Zinc (Zn) Max. 20 ppm
Iron(III) Max. 200 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR