Nitric acid 64 - 66%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger

07006-2.5L
BJ07006-1LEA 53.39 USD
BJ07006-1L BJ07006-2.5L
Nitric acid 64 - 66%, Puriss., Fluka™
Nitric acid
Formula: HNO₃
Boiling Pt: 122 °C (1.013 hPa)
Melting Pt: –31 °C
Density: 1.400…1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Assay 64 - 66 %
Residue on ignition (SO₄) Max. 0.0005 %
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.0005 %
Iron (Fe) Max. 0.0001 %
Vapor Pressure 8 mmHg ( 20 °C)
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR