Potassium hydroxide ≥85%, powder, Technical Grade, Fluka™
Supplier: Honeywell Research Chemicals
06103-5KG
BJ06103-1KGEA
118.23
USD
BJ06103-1KG
BJ06103-5KG
Potassium hydroxide ≥85%, powder, Technical Grade, Fluka™
Potassium hydroxide
Specification Test Results
Assay | Min. 85 % |
Iron (Fe) | Max. 0.002 % |
Chloride (Cl) | Max. 0.1 % |
Sulfate (SO₄) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 0.002 % |
Assay of K₂CO₃ | Max. 1.0 % |
Vapor Pressure | 1 mmHg ( 719 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...