Charcoal activated ≥98.5%, powder, Purum ≤40 μm (75%), Fluka™

Supplier: Honeywell Research Chemicals
Warning

05120-1KG 05120-5KG
BJ05120-1KGEA 187.48 USD
BJ05120-1KG BJ05120-5KG
Charcoal activated ≥98.5%, powder, Purum ≤40 μm (75%), Fluka™
Charcoal activated
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now

Specification Test Results

Loss on drying (105°C) Max. 7 %
Granulation ≤ 40 µm Min.75 % complying
Residue on ignition Max. 7 %
Iron (Fe) Max. 0.3 %
Vapor Pressure <0.1 mmHg ( 20 °C)
Autoignition Temperature 842 °F
Resistivity 1375 μΩ-cm, 20°C (graphite)
Form powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR