Charcoal activated, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Warning

05117-1KG
BJ05117-1KGEA 599.49 USD
BJ05117-1KG
Charcoal activated, tested according to Ph. Eur., Fluka™
Charcoal activated
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now

Specification Test Results

Autoignition Temperature 842 °F
Identity B complying
Alkali soluble colored substances complying
Sulfated ash Max. 5 %
Loss on drying (120°C) Max. 15 %
Acidity or alkalinity complying
Soluble in acid Max. 3 %
Soluble in ethanol Max. 0.5 %
Copper (Cu) Max. 25.0 ppm
Lead (Pb) Max. 10.0 ppm
Zinc (Zn) Max. 25.0 ppm
Sulfide (S) complying
Absorption capacity complying
Fluorescent substances complying
Total bacterial value Max. 1000 KBE/g
Fungi and yeasts Max. 100 KBE/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR