1-Chlorobutane ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

034-4
BJ034-1LCS 2179.2 USD
BJ034-1L BJ034-4
1-Chlorobutane ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
1-Chlorobutane

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃(CH₂)₃Cl
Boiling Pt: 78 °C
Melting Pt: –123 °C
Density: 0.8862 g/cm³ (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity (as HCl) 1 mg/L max.
Chloride 10 mg/L max.
GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.4009 - 1.4025
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.5 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Cutoff 220 nm max.
Water by Karl Fischer Titration 0.02 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR