Sodium iodide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
03129-6X1KG
BJ03129-1KGCS
2281.87
USD
BJ03129-1KG
Sodium iodide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium iodide
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300 Melting Pt: 662 Density: 3.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 Merck Index: 13,08705 |
Specification Test Results
Assay (calculated to the dried substance) | 99.0 - 100.5 % |
Loss on drying (105°C) | Max. 3.0 % |
Water (Karl Fischer) | Max. 2.0 % |
Alkalinely reacting impurities | complying |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | complying |
Heavy metals (as Pb) | Max. 5 ppm |
Iodate (IO₃) | Max. 2 ppm |
Iodate (JO₃) | complying |
Sulfate (SO₄) | Max. 150 ppm |
Thiosulfate (S₂O₃) | Max. 0.005 % |
Thiosulfate and Barium | complying |
Nitrate, nitrite, ammonium | complying |
Residual solvents | complying |
Appearance of the solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...