Ammonium iodide ≥99%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals

03101-6X1KG
BJ03101-1KGCS 3120.92 USD
BJ03101-1KG
Ammonium iodide ≥99%, Puriss., Fluka™
Ammonium iodide
Formula: NH₄I
MW: 144.94 g/mol
Density: 2.510 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7

Order Now

Specification Test Results

Assay Min. 99.0 %
Loss on drying (105°C) Max. 1 %
Residue on ignition (as sulfate) Max. 0.1 %
pH (5 %, 20°C) 4.5 - 6.5
Arsenic (As) Max. 0.0005 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride and bromide (as Cl) Max. 0.02 %
Iodate (IO₃) Max. 0.01 %
Sulfate (SO₄) Max. 0.01 %
Thiosulfate (S₂O₃) Max. 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR