Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
01379-1L
BJ01379-1LEA
260.86
USD
BJ01379-1L
Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Heptane (mixture of isomers)
High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.
Formula:
H₃C(CH₂)₅CH₃ MW: 100.2 g/mol Boiling Pt: 98.2…98.4 °C (1.000 hPa) Melting Pt: –90.5 °C Density: 0.69 g/cm³ (15 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II |
|
Specification Test Results
Assay (GC) | Min. 99.8 % |
Water (Karl Fischer) | Max. 0.005 % |
Silver (Ag) | Max. 0.5 ppb |
Aluminium (Al) | Max. 1 ppb |
Arsenic (As) | Max. 1 ppb |
Gold (Au) | Max. 0.5 ppb |
Barium (Ba) | Max. 0.5 ppb |
Beryllium (Be) | Max. 0.5 ppb |
Bismuth (Bi) | Max. 0.5 ppb |
Calcium (Ca) | Max. 20 ppb |
Cadmium (Cd) | Max. 0.5 ppb |
Cerium (Ce) | Max. 0.5 ppb |
Cobalt (Co) | Max. 0.5 ppb |
Chromium (Cr) | Max. 1 ppb |
Cesium (Cs) | Max. 0.5 ppb |
Copper (Cu) | Max. 1 ppb |
Iron (Fe) | Max. 1 ppb |
Gallium (Ga) | Max. 0.5 ppb |
Germanium (Ge) | Max. 0.5 ppb |
Mercury (Hg) | Max. 1 ppb |
Indium (In) | Max. 0.5 ppb |
Iridium (Ir) | Max. 0.5 ppb |
Potassium (K) | Max. 10 ppb |
Lithium (Li) | Max. 0.5 ppb |
Magnesium (Mg) | Max. 1 ppb |
Manganese (Mn) | Max. 0.5 ppb |
Molybdenum (Mo) | Max. 0.5 ppb |
Sodium (Na) | Max. 100 ppb |
Nickel (Ni) | Max. 1 ppb |
Lead (Pb) | Max. 0.5 ppb |
Palladium (Pd) | Max. 0.5 ppb |
Platinum (Pt) | Max. 0.5 ppb |
Rubidium (Rb) | Max. 0.5 ppb |
Rhodium (Rh) | Max. 0.5 ppb |
Ruthenium (Ru) | Max. 0.5 ppb |
Antimony (Sb) | Max. 0.5 ppb |
Selenium (Se) | Max. 1 ppb |
Tin (Sn) | Max. 0.5 ppb |
Strontium (Sr) | Max. 0.5 ppb |
Titanium (Ti) | Max. 1 ppb |
Thallium (Tl) | Max. 0.5 ppb |
Vanadium (V) | Max. 0.5 ppb |
Zinc (Zn) | Max. 1 ppb |
Zirconium (Zr) | Max. 0.5 ppb |
Refractive Index | n20/D 1.387 (lit.) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...