Sulfuric acid 0.25 M H2SO4 (0.5N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
35355-10L-VP
35355-1L
BJ35355-10L-VPEA
300.16
USD
BJ35355-10L-VP
BJ-35355-1L
Sulfuric acid 0.25 M H2SO4 (0.5N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.020 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 203-755-4 UN: 3264 ADR: 8 III |
Specification Test Results
Factor | 0.999 - 1.001 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...