Sodium hydroxide 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

35263-10L-VP 35263-6X1L 35263-5L-VP 35263-5L
BJ35263-10L-VPEA 277.02 USD
BJ35263-10L-VP BJ35263-1L BJ35263-5L-VP BJ-35263-5L
Sodium hydroxide 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR