Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

35257-10L-VP 35257-6X1L 35257-5L
BJ35257-10L-VPEA 499 USD
BJ35257-10L-VP BJ35257-1L BJ35257-5L-VP BJ-35257-5L
Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: >100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.020 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR