EDTA disodium salt 0,0178 mol/l (N/28; 1 ml = 1°dH/100 ml H₂O) in aqueous solution volumetric solution, for hardness of water determinations, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

34543-1L
BJ-34543-1LEA 151.74 USD
BJ-34543-1L
EDTA disodium salt 0,0178 mol/l (N/28; 1 ml = 1°dH/100 ml H₂O) in aqueous solution volumetric solution, for hardness of water determinations, Fluka™
EDTA disodium salt

1 ml = 1 German degree of hardness in 100 ml of water

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 231-791-2
Merck Index: 12,03556

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR