N,N-Dimethylformamide ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34489-2.5L
BJ34489-4LCS 3329.16 USD
BJ34489-4L BJ-34489-2.5L
N,N-Dimethylformamide ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
N,N-Dimethylformamide

Dimethylformamide is an organic compound with the formula (CH3)2NC(O)H, commonly abbreviated as DMF. This colourless liquid is miscible with water and many hydrophobic organic compounds and has a low evaporation rate. Suitable for Pesticide Residue Aanlysis (PRA).


  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: HCON(CH₃)₂
Boiling Pt: 152…153 °C (1.013 hPa)
Melting Pt: –61 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) min. 99.8 %
Water (Karl Fischer) max. 0.08 %
Non-volatile matter max. 0.001 %
Suitability for residue analysis max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR