Biofit Ergonomic Lab Bench Chairs
Supplier: BioFit
AEW-L-RC-T-XF-XA-C-P28540
BE-AEP1722RCEA 1670.34 USD
BE-AEP1722RC
Biofit Ergonomic Lab Bench Chairs
Chairs
BioFit chairs offer an excellent combination of durability, comfort, and ergonomic support for lab facilities and other workplaces. The chairs have many ergonomic features that help users remain fresh and productive over long work sessions.
  • Sculptured, saddle-shaped seat with waterfall front improves circulation and reduces strain on the lower legs
  • Generous ergonomic backrest provides lumbar support for the lower back
  • Black vinyl seat, casters, and chrome finish
The high bench chair has an adjustable footring. The low bench chair and desk chair do not have a footring.