Sodium metabisulfite ≥97.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

BDH9282-500G
BDH9316-500GCS 685.81 USD
BDH9316-500G
Sodium metabisulfite ≥97.0% ACS, VWR Chemicals BDH®
Sodium metabisulphite
Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride =0.05%
Heavy Metals (as Pb) =0.001%
Insolubles =0.005%
Iron =0.002%
Assay = 97.0 %
Thiosulfate =0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR