Zinc chloride ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

BDH9314-500G
BDH9314-500GCS 3957.35 USD
BDH9314-500G
Zinc chloride ACS analytical reagent, VWR Chemicals BDH®
Zinc chloride

White free-flowing granules.


  • Assay percent range of greater than 97%
  • Certified ACS reagent grade
  • Available in a variety of volumes


Zinc chloride is available as white to off-white crystalline powder, crystals, or chunks.

Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

Ammonium = 0.005 %
Calcium = 0.06 %
Insolubles = 0.005 %
Iron = 0.001 %
Lead = 0.005 %
Magnesium = 0.01 %
Nitrate = 0.003 %
Oxychloride PASS
Potassium = 0.02 %
Purity = 97.0 %
Sodium = 0.05 %
Sulfate (SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR