Trichloroacetic acid (TCA) ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

BDH9310-500G
BDH9310-500GCS 2452.42 USD
BDH9310-500G
Trichloroacetic acid (TCA) ACS analytical reagent, VWR Chemicals BDH®
Trichloroacetic acid (TCA)

White or nearly colorless moist crystal plates. Soluble in water.

Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.002 %
Clarity of Solution PASS
Heavy Metals = 0.002 %
Insoluble Matter = 0.01 %
Iron = 0.001 %
Nitrate = 0.002 %
Phosphate = 5 ppm
Purity = 99.0 %
Residue after Ignition = 0.03 %
Substances Darkened by H2SO4 PASS
Sulfate = 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR