Tin(II) chloride dihydrate 98.0-103.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Stannous chloride dihydrate

BDH9304-500G
BDH9304-500GCS 3195.24 USD
BDH9304-500G
Tin(II) chloride dihydrate 98.0-103.0% ACS, VWR Chemicals BDH®
Tin (II) chloride dihydrate

White to off-white solid

Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay 98.0 - 103.0 %
Calcium <= 0.005 %
Iron <= 0.003 %
Lead <= 0.01 %
Potassium <= 0.005 %
Sodium <= 0.01 %
Solubility in hydrochloric acid PASS
Sulfate (P/F) PASS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR