Sodium fluoride ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

0581VBD500G
BDH9290-500GCS 1515.37 USD
BDH9290-500G
Sodium fluoride ≥99.0% ACS, VWR Chemicals BDH®
Sodium fluoride

Meets reagent specifications for testing USP/NF monographs.


White solid

Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.005 %
Heavy Metals (as Pb) = 0.003 %
Insolubles = 0.02 %
Iron = 0.003 %
Loss on Drying = 0.3 %
Potassium = 0.02 %
Purity = 99.0 %
Sodium Fluosilicate = 0.1 %
Sulfate = 0.03 %
Sulfite = 0.005 %
Titrable Free Acid = 0.03 meq/g
Titrable Free Base = 0.01 meq/g
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR