tri-Sodium citrate dihydrate ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

0101VBD2.5KG
BDH9288-2.5KGCS 786.92 USD
BDH9288-2.5KG BDH9288-500G
tri-Sodium citrate dihydrate ≥99.0% ACS, VWR Chemicals BDH®
tri-Sodium citrate dihydrate

Colorless or white solid

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonia = 0.003 %
Calcium = 0.005 %
Chloride = 0.003 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
pH (5%, Water) @25C 7.0 - 9.0
Purity = 99.0 %
Sulfate = 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR