Sodium chloride ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Saline, NaCl, Sea salt, Halite

0241VBD2.5KG BDH9286-12KG BDHVBDH9286-250LB 0241VBD500G BDH9286-50KG
BDH9286-2.5KGCS 381.73 USD
BDH9286-2.5KG BDH9286-12KG BDH9286-250LB BDH9286-500G BDH9286-50KG
Sodium chloride ≥99.0% ACS, VWR Chemicals BDH®
Sodium chloride

Meets reagent specifications for testing USP/NF monographs.


White solid

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Barium Test PASS
Bromide <= 0.01 %
Calcium <= 0.002 %
Chlorate & Nitrate <= 0.003 %
Heavy Metals <= 0.0005 %
Insolubles <= 0.005 %
Iodide <= 0.002 %
Iron <= 0.0002 %
Magnesium <= 0.001 %
pH (5%, Water) @25C 5.0 - 9.0
Phosphate <= 0.0005 %
Potassium <= 0.005 %
Purity >= 99.0 %
Sulfate <= 0.004 %
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR