Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

BDH9284-2.5KG BDH9284-500G
BDH9284-2.5KGCS 677.84 USD
BDH9284-2.5KG BDH9284-500G
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Sodium carbonate

Meets reagent specifications for testing USP/NF monographs.


White granular powder.

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <= 0.03 %
Chloride <= 0.001 %
Heavy Metals (as Pb) <= 0.0005 %
Insolubles <= 0.01 %
Iron <= 0.0005 %
Loss on Heating <= 1.0 %
Magnesium <= 0.005 %
Phosphate <= 0.001 %
Potassium <= 0.005 %
Purity >= 99.5 %
Silica <= 0.005 %
Sulfur Compounds <= 0.003 %
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR