Sodium hydrogen carbonate 99.7-100.3% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Sodium bicarbonate

BDH9280-12KG 0865VBD500G BDH9280-2.5KG
BDH9280-12KGEA 251.41 USD
BDH9280-12KG BDH9280-500G BDH9280-2.5KG
Sodium hydrogen carbonate 99.7-100.3% ACS, VWR Chemicals BDH®
Sodium hydrogen carbonate

Meets reagent specifications for testing USP/NF monographs.


White solid

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium = 0.0005 %
Calcium = 0.02 %
Chloride = 0.003 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.015 %
Iron = 0.001 %
Magnesium = 0.005 %
Phosphate = 0.001 %
Potassium = 0.005 %
Purity 99.7 - 100.3 %
Sulfur Compounds = 0.003 %
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR