Silver nitrate ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH9276-125G BDH9276-500G BDH9276500G
BDH9276-125GCS 10372.81 USD
BDH9276-125G BDH9276-500G BDH9276500G
Silver nitrate ACS analytical reagent, VWR Chemicals BDH®
Silver nitrate

Colorless or white solid

Formula: AgNO₃
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless or white solid/crystals
Chloride <= 0.0005 %
Clarity of Solution PASS
Copper <= 0.0002 %
Free Acid Test PASS
Iron <= 0.0002 %
Lead <= 0.001 %
Purity >= 99.0 %
Substances not Precipitated by HCl <= 0.01 %
Sulfate <= 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR