ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

BDH9266-500G BDH9266-2.5KG
BDH9266-500GCS 1456.29 USD
BDH9266-500G BDH9266-2.5KG
ACS analytical reagent, VWR Chemicals BDH®
di-Potassium hydrogen phosphate

White solid.

Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.003 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.01 %
Iron = 0.001 %
Loss on Drying = 1.0 %
Nitrogen Compounds = 0.001 %
pH (5%, Water) @25C 8.5 - 9.6
Purity = 98.0 %
Sodium = 0.05 %
Sulfate = 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR