Potassium iodide ACS, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Dipotassium diiodide, Kali iodide
BDH9264-2.5KG
BDH9264-125G
BDH9264-500G
BDH9264-2.5KGCS
3981.3
USD
BDH9264-2.5KG
BDH9264-125G
BDH9264-500G
Potassium iodide ACS, VWR Chemicals BDH®
Potassium iodide
Meets reagent specifications for testing USP/NF monographs.
Colorless or white solid
Formula:
Kl MW: 166 g/mol Boiling Pt: 1330 °C (1013 hPa) Melting Pt: 686 °C Density: 3.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 Merck Index: 13,07727 |
Specification Test Results
Barium | <= 0.002 % |
Calcium | <= 0.002 % |
Chloride & Bromide | <= 0.01 % |
Heavy Metals (as Pb) | <= 0.0005 % |
Insolubles | <= 0.005 % |
Iodate | <= 0.0003 % |
Iron | <= 0.0003 % |
Loss on Drying | <= 0.2 % |
Magnesium | <= 0.001 % |
pH (5%, Water) @25C | 6.0 - 9.2 |
Phosphate | <= 0.001 % |
Purity | >= 99.0 % |
Sodium | <= 0.005 % |
Sulfate | <= 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...