Potassium carbonate ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDH9256-500G
BDH9256-500GCS 939.56 USD
BDH9256-500G
Potassium carbonate ≥99.0% ACS, VWR Chemicals BDH®
Potassium carbonate

Meets reagent specifications for testing USP/NF monographs.


White, hygroscopic solid

Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity = 99.0 %
Insoluble matter = 0.01 %
Chloride (Cl) = 0.003 %
Phosphate (PO4) = 0.001 %
Silica (SiO2) = 0.005 %
Sulfur compounds (as SO4) = 0.004 %
Heavy metals (as Pb) = 0.0005 %
Iron (Fe) = 0.0005 %
Calcium (Ca) = 0.005 %
Magnesium (Mg) = 0.002 %
Sodium (Na) = 0.02 %
Mets reagent specifications for testing USP/NF monographs.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR