Potassium acetate ≥99.0% ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: 4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid, Diuretic salt, Acetic acid potassium salt, Acetic acid potassium salt

BDH9254-500G
BDH9254-500GCS 985.37 USD
BDH9254-500G
Potassium acetate ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Potassium acetate

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid

Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium = 0.005 %
Chloride = 0.003 %
Heavy Metals = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
Magnesium = 0.002 %
pH (5%, Water) @25C 6.5 - 9.0
Phosphate = 0.001 %
Purity = 99.0 %
Sodium = 0.03 %
Sulfate = 0.002 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR