D-(-)-Mannitol ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

BDH9248-500G
BDH9248-500GCS 1220.75 USD
BDH9248-500G
D-(-)-Mannitol ACS, VWR Chemicals BDH®
D(-)-Mannitol

White solid.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals = 0.0005 %
Insoluble Matter = 0.01 %
Loss on Drying = 0.05 %
Reducing Sugars Test PASS
Residue after Ignition = 0.01 %
Specific Rotation Test +23.3 - +24.3 degrees
Titrable Acid = 0.0008 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR