Magnesium sulfate ≥98.0% ACS, Reagent Grade analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous
BDH9246-500G
BDH9246-500GCS
1093.24
USD
BDH9246-500G
Magnesium sulfate ≥98.0% ACS, Reagent Grade analytical reagent, VWR Chemicals BDH®
Magnesium sulphate
White powder.
Formula:
MgSO₄ MW: 120.37 g/mol Melting Pt: 1127 Density: 2.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011110 CAS Number: 7487-88-9 EINECS: 231-298-2 Merck Index: 12,05731 |
|
Specification Test Results
Purity | ≥98.0 % |
Moisture | ≤0.2 % |
Appearance | Fine white to off-white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...