Magnesium chloride hexahydrate 99.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
BDH9244-12KG
BDH9244-2.5KG
BDH9244-500G
BDH9244-12KGEA
427.54
USD
BDH9244-12KG
BDH9244-2.5KG
BDH9244-500G
Magnesium chloride hexahydrate 99.0-102.0% ACS, VWR Chemicals BDH®
Magnesium chloride hexahydrate
Meets reagent specifications for testing USP/NF monographs.
Colorless, deliuescent solid
Formula:
MgCl₂·6H₂O Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C Density: 1.57 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Purity | 99.0-102.0 % |
Ammonium | ≤0.002 % |
Barium | ≤0.005 % |
Calcium | ≤0.01 % |
Heavy Metals (as Pb) | ≤0.0005 % |
Insolubles | ≤0.005 % |
Iron | ≤0.0005 % |
Manganese | ≤0.0005 % |
Nitrate | ≤0.001 % |
Phosphate | ≤0.0005 % |
Potassium | ≤0.005 % |
Sodium | ≤0.005 % |
Strontium | ≤0.005 % |
Sulfate | ≤0.002 % |
Meets reagent specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...