Hydroxylammonium chloride ≥96.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

BDH9236-1KG BDH9236-500G
BDH9236-1KGCS 1740.77 USD
BDH9236-1KG BDH9236-500G
Hydroxylammonium chloride ≥96.0% ACS, VWR Chemicals BDH®
Hydroxylammonium chloride

Meets reagent specifications for testing USP/NF monographs.


Colorless, hygroscopic solid

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Ammonium (P/F) PASS
Clarity of Alcohol Solution PASS
Heavy Metals = 0.0005 %
Iron = 0.0005 %
Purity = 96.0 %
Residue on Ignition = 0.05 %
Sulfur Compounds = 0.005 %
Titrable Free Acid = 0.25 meq/g
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR