EDTA (ethylenediamine tetraacetic acid) ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, Edetic Acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, EDTA, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

BDH9232-500G
BDH9232-500GCS 2388.7 USD
BDH9232-500G
EDTA (ethylenediamine tetraacetic acid) ACS, VWR Chemicals BDH®
EDTA (Ethylenediamine tetraacetic acid)

White solid.

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium = 0.001 %
Heavy Metals (as Pb) = 0.001 %
Insoluble in Dilute NH4OH = 0.005 %
Iron = 0.005 %
Magnesium = 0.0005 %
Nitrilotriacetic Acid = 0.1 %
Purity 99.4 - 100.6 %
Residue after Ignition = 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR