D-(+)-Glucose, anhydrous ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

BDH9230-500G BDH9230-2.5KG
BDH9230-500GCS 740.25 USD
BDH9230-500G BDH9230-2.5KG
D-(+)-Glucose, anhydrous ACS, VWR Chemicals BDH®
D(+)-Glucose

Meets reagent specifications for testing USP/NF monographs.


White solid

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.01 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
Loss on Drying = 0.2 %
Residue on Ignition = 0.02 %
Specific Rotation +52.5 - +53.0 degrees
Starch PASS
Sulfate & Sulfite = 0.005 %
Titrable Acid = 0.002 meq/g
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR