Calcium nitrate tetrahydrate ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH9226-2.5KG BDH9226-500G
BDH9226-2.5KGCS 1804.07 USD
BDH9226-2.5KG BDH9226-500G
Calcium nitrate tetrahydrate ACS analytical reagent, VWR Chemicals BDH®
Calcium nitrate tetrahydrate

Colorless, hygroscopic solid

Formula: Ca(NO₃)₂·4H₂O
Melting Pt: ∼42 °C
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 13,01687

Order Now

Specification Test Results

Barium = 0.005 %
Chloride = 0.005 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
Magnesium = 0.05 %
Nitrite = 0.001 %
pH (5%, Water) @25C 5.0 - 7.0
Potassium = 0.005 %
Purity 99.0 - 103.0 %
Sodium = 0.01 %
Strontium = 0.05 %
Sulfate = 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR