Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDH9210-500G
BDH9210-500GCS 3131.16 USD
BDH9210-500G
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS analytical reagent, VWR Chemicals BDH®
Ammonium heptamolybdate tetrahydrate

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid; sometimes slight green/yellow tint

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Arsenate, Phosphate and Silicate = 0.001 %
Chloride = 0.002 %
Heavy Metals (as Pb) = 0.001 %
Insolubles = 0.005 %
Magnesium = 0.005 %
MoO3 81.0 - 83.0 %
Nitrate = 0.003 %
Phosphate = 0.0005 %
Potassium = 0.01 %
Sodium = 0.01 %
Sulfate = 0.02 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR