Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®

Supplier: VWR International

BDH85800.100E BDH85800.400 BDH85800.201 BDH85800.202 BDH85800.050 BDH85800.100
BDH85800.100ECS 380.57 USD
BDH85800.100E BDH85800.400 BDH85800.201 BDH85800.202 BDH85800.050 BDH85800.100
Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
Methanol

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


Solvents are pumped through 0.2micron filters on filling and packaged under nitrogen.


Packaging: All bottles have tamper evident caps for added peace of mind and are packed in environmentally –friendly corrugated/pulp case inserts that are fully recyclable.

Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% Min.
Appearance Clear colourless liquid
Carbonyl compounds Passes test
Solubility in water Passes test
Substances darkened by sulphuric acid Passes test
Substances reducing permanganate Passes test
Acidity 0.0002 meq/g Max.
Alkalinity 0.0002 meq/g Max.
Boiling point 64 - 65 °C
Colouration 10 APHA Max.
Density (20/20) 0.791 - 0.793
Evaporation residue 1 ppm Max.
Water 0.02 % Max.
Al (Aluminium) 20 ppb Max
Ag (Silver) 100 ppb Max
As (Arsenic) 20 ppb Max
Ba (Barium) 100 ppb Max
Ca (Calcium) 200 ppb Max
Cd (Cadmium) 20 ppb Max
Co (Cobalt) 20 ppb Max
Cr (Chromium) 20 ppb Max
Cu (Copper) 10 ppb Max
Fe (Iron) 50 ppb Max
K (Potassium) 50 ppb Max
Mg (Magnesium) 50 ppb Max
Mn (Manganese) 100 ppb Max
Mo (Molybdenum) 20 ppb Max
Na (Sodium) 200 ppb Max
Ni (Nickel) 100 ppb Max
Pb (Lead) 20 ppb Max
V (Vanadium) 20 ppb Max
Zn (Zinc) 100 ppb Max
Gradient (220 nm) 3 mAU Max
Gradient (235 nm) 2 mAU Max
Gradient (254 nm) 1 mAU Max
Transmittance (210 nm) 45% Min
Transmittance (220 nm) 65% Min
Transmittance (225 nm) 70% Min
Transmittance (230 nm) 85% Min
Transmittance (235 nm) 90% Min
Transmittance (240 nm) 95% Min
Transmittance (250 nm) 95% Min
Transmittance (260 nm) 98% Min
Transmittance (280-400 nm) 98% Min
Absorbance (210 nm) 0.347 Max
Absorbance (220 nm) 0.188 Max
Absorbance (225 nm) 0.155 Max
Absorbance (230 nm) 0.071 Max
Absorbance (235 nm) 0.046 Max
Absorbance (240 nm) 0.023 Max
Absorbance (250 nm) 0.023 Max
Absorbance (260 nm) 0.009 Max
Absorbance (280-400 nm) 0.009 Max
Fluorescence (as quinine) (254 nm) 1 ppb Max
Fluorescence (as quinine) (365 nm) 0.5 ppb Max
MS-ESI+ (as Reserpine) 50 ppb Max
MS-APCI+ (as Reserpine) 50 ppb Max
MS-ESI- (as 4-Nitrophenol) 20 ppb Max
MS-APCI- (as 4-Nitrophenol) 20 ppb Max
Conforms Ph.Eur. R1 1053201 Passes test
Conforms Ph.Eur. R2 1053202 Passes test
Conforms to ACS Passes test
Filtered at 0.2 μm Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR