Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Supplier: VWR International
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
- Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.