Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS

Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

BDH83640.400CS 1214.84 USD
BDH83640.400 BDH83640.100E
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Acetonitrile

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


Solvents are pumped through 0.2 micron filters on filling and packaged under nitrogen.


Ordering information: To purchase these solvents in stainless steel returnable drums as part of the VWR ReCAP program, please contact your local VWR sales representative. A separate contract for VWR ReCAP products is required.


Packaging: All bottles have tamper evident caps for added peace of mind and are packed in environmentally –friendly corrugated/pulp case inserts that are fully recyclable.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0001 mEq/g
Alkalinity Max. 0.0002 meq/g
Coloration Max. 10 APHA
Density (20/4) 0.781-0.784
n 20/D 1.343-1.345
Evaporation residue Max. 2 ppm
Water Max. 0.02 %
Al (Aluminium) Max. 50 ppb
Ca (Calcium) Max. 50 ppb
Fe (Iron) Max. 50 ppb
K (Potassium) Max. 50 ppb
Mg (Magnesium) Max. 10 ppb
Na (Sodium) Max. 100 ppb
Transmittance (191 nm) Min. 30 %
Transmittance (195 nm) Min. 80 %
Transmittance (200 nm) Min. 95 %
Transmittance (210 nm) Min. 96 %
Transmittance (215 nm) Min. 98 %
Transmittance (220 nm) Min. 98 %
Transmittance (230 nm) Min. 99 %
Transmittance (from 240 nm) Min. 99 %
Fluorescence (as quinine) (254/450 nm) Max. 1 ppb
Gradient grade (210 nm) Max. 3 mAU
Gradient grade (254 nm) Max. 1 mAU
Mode: ESI 200 µl pos | APCI 200 µl pos Max. 2 ppb
Mode: ESI 200 µl neg | APCI 200 µl Max. 20 ppb
Conforms to Reag. Ph.Eur. Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR